(6-Bromo-2-pyridinyl)methanol 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(6-Bromopyridin-2-yl)methanol, 2-Bromo-6-pyridinemethanol, 2-Bromo-6-(hydroxymethyl)pyridine
H56123-03
H56123-06
H56123-MD
CAAAH56123-03EA
100.01
CAD
CAAAH56123-03
CAAAH56123-06
CAAAH56123-MD
(6-Bromo-2-pyridinyl)methanol 96%
(6-Bromo-2-pyridinyl)methanol
CAS: 33674-96-3
MDL: MFCD04039884
Formula:
C₆H₆BrNO MW: 188.02 g/mol Boiling Pt: 246 °C Melting Pt: 34…39 °C Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD04039884 CAS Number: 33674-96-3 |
Specification Test Results
Assay Minimum | 96 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...