1,1,4,7,10,10-Hexamethyltriethylenetetramine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H56089-03
H56089-06
H56089-14
CAAAH56089-03EA
54.35
CAD
CAAAH56089-03
CAAAH56089-06
CAAAH56089-14
1,1,4,7,10,10-Hexamethyltriethylenetetramine 97%
1,1,4,7,10,10-Hexamethyltriethylenetetramine
Purity: 97%
CAS No.: 3083-10-1
Molecular Formula: C12H30N4
Formula Weight: 230.39
Flash Point: 102°(216°F)
Refractive Index: 1.4560
Boiling Point: 130°/11mm
UN No.: 3725
MDL No.: MFCD00025684
Formula:
C₁₂H₃₀N₄ Boiling Pt: 130 °C/11 mm Density: 0.847 Flash Pt: 102 °C (216 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00025684 CAS Number: 3083-10-1 EINECS: 221-382-7 UN: 2735 ADR: 8,III |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...