(R)-(-)-2,5-Dihydro-3,6-dimethoxy-2-isopropylpyrazine 98%
Supplier: Thermo Scientific Chemicals
Warning
|
H56077-AA
H56077-XF
CAAAH56077-AAEA
801.06
CAD
CAAAH56077-AA
CAAAH56077-XF
(R)-(-)-2,5-Dihydro-3,6-dimethoxy-2-isopropylpyrazine 98%
(R)-(-)-2,5-Dihydro-3,6-dimethoxy-2-isopropylpyrazine
CAS: 109838-85-9
MDL: MFCD00040565
Formula:
C₉H₁₆N₂O₂ Density: 1.028 Flash Pt: 85 °C(185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00040565 CAS Number: 109838-85-9 |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...