3-Methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate ≥98% (by HPLC)

Supplier: Thermo Scientific Chemicals
Danger

H56061-06 H56061-18
CAAAH56061-06EA 97.17 CAD
CAAAH56061-06 CAAAH56061-18
3-Methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate ≥98% (by HPLC)
3-Methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate

It is used as a reagent for the spectrophotometric determination of aliphatic aldehydes and for the determination of hexosamines in glycosaminoglycans. It is Incorporated into a new peroxidase color reaction. It is also a reagent for the spectrophotometric determination of traces of selenium (IV) in environmental samples.


Caution: Incompatible with oxidizing agents. Store at room temperature.

Formula: C₈H₉N₃S·H₂O·HCl
Melting Pt: 270…274 °C
Storage Temperature: Ambient
MDL Number: MFCD00149370
CAS Number: 38894-11-0
EINECS: 238-428-7
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay (HPLC) 98.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR