6-Bromooxindole 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Bromo-2-oxindole, 6-Bromoindolin-2-one

H56012-03 H56012-06
CAAAH56012-03EA 73.63 CAD
CAAAH56012-03 CAAAH56012-06
6-Bromooxindole 97%
6-Bromooxindole

CAS: 99365-40-9 MDL: MFCD02179605

Formula: C₈H₆BrNO
MW: 212.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02179605
CAS Number: 99365-40-9

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR