Iodine trichloride (Wijs'reagent) 97%

Supplier: Thermo Scientific Chemicals
Danger

H56005-06 H56005-14 H56005-22
CAAAH56005-06EA 61.46 CAD
CAAAH56005-06 CAAAH56005-14 CAAAH56005-22
Iodine trichloride (Wijs'reagent) 97%
Iodine trichloride (Wijs'reagent)

CAS: 865-44-1 MDL: MFCD00036291

Formula: ICl₃
Melting Pt: 63 °C
Density: 1.23 g/cm³ (20 °C)
Flash Pt: 40 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00036291
CAS Number: 865-44-1
EINECS: 212-739-8
UN: 3260
ADR: 8,III
Merck Index: 14,5020

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR