6-Hydroxyindole 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H55951-03
H55951-06
H55951-MD
CAAAH55951-03EA
270.18
CAD
CAAAH55951-03
CAAAH55951-06
CAAAH55951-MD
6-Hydroxyindole 98%
6-Hydroxyindole
CAS: 2380-86-1
MDL: MFCD00152101
Formula:
C₈H₇NO Melting Pt: 125…128 °C Storage Temperature: Ambient |
MDL Number:
MFCD00152101 CAS Number: 2380-86-1 EINECS: 417-020-4 UN: 3077 ADR: 9,III |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...