1-Benzylpyrrole 96%

Supplier: Thermo Scientific Chemicals
Warning

H55936-03 H55936-06 H55936-14
CAAAH55936-03EA 48.98 CAD
CAAAH55936-03 CAAAH55936-06 CAAAH55936-14
1-Benzylpyrrole 96%
1-Benzylpyrrole

Purity: 97% CAS No.: 2051-97-0 Molecular Formula: C11H11N Formula Weight: 157.21 Physical Form: Liquid Density: 1.020 Refractive Index: 1.5685 Appearance: Colorless to brown Boiling Point: 90°/0.9mm UN No.: 1993 MDL No.: MFCD00963301

Formula: C₁₁H₁₁N
MW: 157.22 g/mol
Boiling Pt: 90 °C/0.9 mm
Density: 1.020
Storage Temperature: Ambient
MDL Number: MFCD00963301
CAS Number: 2051-97-0
EINECS: 218-137-1
UN: 1993
ADR: "3",III

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR