2-(Tributylstannyl)thiophene 97%

Supplier: Thermo Scientific Chemicals
Danger

H55904-AA H55904-AC H55904-AE
CAAAH55904-AAEA 70.15 CAD
CAAAH55904-AA CAAAH55904-AC CAAAH55904-AE
2-(Tributylstannyl)thiophene 97%
2-(Tributylstannyl)thiophene

CAS: 54663-78-4 MDL: MFCD00192513

Formula: C₁₆H₃₀SSn
Boiling Pt: 155 °C/0.1 mm
Density: 1.175
Storage Temperature: Ambient
MDL Number: MFCD00192513
CAS Number: 54663-78-4
UN: 2788
ADR: "6.1",III

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR