3-Hexylthiophene 99+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H55891-03
H55891-06
H55891-14
CAAAH55891-03EA
53.25
CAD
CAAAH55891-03
CAAAH55891-06
CAAAH55891-14
3-Hexylthiophene 99+%
3-Hexylthiophene
CAS: 1693-86-3
MDL: MFCD00143181
Formula:
C₁₀H₁₆S Boiling Pt: 65 °C/0.45 mm Density: 0.936 Flash Pt: 37 °C (99 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00143181 CAS Number: 1693-86-3 UN: 1993 ADR: 3,III |
Specification Test Results
Assay Minimum | 99 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...