4-Methoxybenzenediazonium tetrafluoroborate 98%

Supplier: Thermo Scientific Chemicals
Danger

H55827-03 H55827-06 H55827-14
CAAAH55827-03EA 53.4 CAD
CAAAH55827-03 CAAAH55827-06 CAAAH55827-14
4-Methoxybenzenediazonium tetrafluoroborate 98%
4-Methoxybenzenediazonium tetrafluoroborate

Purity: 98% CAS No.: 459-64-3 Molecular Formula: C7H7BF4N2O Formula Weight: 221.95 Melting Point: 142-144° UN No.: 3261 MDL No.: MFCD00011897

Formula: C₇H₇BF₄N₂O
MW: 221.95 g/mol
Melting Pt: 142…144 °C
Storage Temperature: Ambient
MDL Number: MFCD00011897
CAS Number: 459-64-3
EINECS: 207-296-2
UN: 2923
ADR: 8,III

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR