2-Nitrofluorene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Nitro-9H-fluorene

H55744-06 H55744-14
CAAAH55744-06EA 58.78 CAD
CAAAH55744-06 CAAAH55744-14
2-Nitrofluorene 98%
2-Nitrofluorene

CAS: 607-57-8 MDL: MFCD00001117

Formula: C₁₃H₉NO₂
MW: 211.22 g/mol
Melting Pt: 156…158 °C
Storage Temperature: Ambient
MDL Number: MFCD00001117
CAS Number: 607-57-8
EINECS: 210-138-5
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR