Azidotri-n-butyltin(IV) 95%

Supplier: Thermo Scientific Chemicals
Danger

H55634-03 H55634-06
CAAAH55634-03EA 49.14 CAD
CAAAH55634-03 CAAAH55634-06 CAAAH55634-14
Azidotri-n-butyltin(IV) 95%
Azidotri-n-butyltin(IV)

Purity: 95% CAS No.: 17846-68-3 Molecular Formula: C12H27N3Sn Formula Weight: 332.06 Flash Point: 113°(235°F) UN No.: 2788 MDL No.: MFCD00216557

Formula: C₁₂H₂₇N₃Sn
Density: 1.212
Flash Pt: 113 °C(235 °F)
Storage Temperature: Ambient
MDL Number: MFCD00216557
CAS Number: 17846-68-3
UN: 2788
ADR: "6.1",III

Order Now

Specification Test Results

Assay Minimum 95


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR