2,4-Dichloro-5-methylpyrimidine 98%

Supplier: Thermo Scientific Chemicals
Danger

H55206-03 H55206-06
CAAAH55206-03EA 79.79 CAD
CAAAH55206-03 CAAAH55206-06 CAAAH55206-MD
2,4-Dichloro-5-methylpyrimidine 98%
2,4-Dichloro-5-methylpyrimidine

CAS: 1780-31-0 MDL: MFCD00023197

Formula: C₅H₄Cl₂N₂
Boiling Pt: 108…109 °C/11 mm
Melting Pt: 26…28 °C
Density: 1.39
Flash Pt: 113 °C (235 °F)
Storage Temperature: Ambient
MDL Number: MFCD00023197
CAS Number: 1780-31-0
EINECS: 217-227-8
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR