2-(Trimethylsilyl)ethoxymethyl chloride tech. 90% stabilised

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-(Chloromethoxy)ethyltrimethylsilane, 2-(Trimethylsilyl)ethoxymethyl Chloride, SEM-chloride

Stabilization: 0.1% Stabilised with N-ethyldiisopropylamine (N,N-diisopropylethylamine)

H55074-03 H55074-06 H55074-14
CAAAH55074-03EA 74.58 CAD
CAAAH55074-03 CAAAH55074-06 CAAAH55074-14
2-(Trimethylsilyl)ethoxymethyl chloride tech. 90% stabilised
2-(Trimethylsilyl)ethoxymethyl chloride

2-(Chloromethoxy)ethyltrimethylsilane is used in the preparation of SEM [2-(trimethylsilyl)ethoxy]methyl]-ethers. It serves as a phenol protecting group in the synthesis of laterifluorones. Further, it reacts with 1H-imidazole to prepare 1-(2-trimethylsilanyl-ethoxymethyl)-1H-imidazole.


CAS: 76513-69-4 MDL: MFCD00009919

Formula: C₆H₁₅ClOSi
MW: 166.72 g/mol
Boiling Pt: 170…172 °C
Density: 0.942
Flash Pt: 46 °C (115 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009919
CAS Number: 76513-69-4
EINECS: 278-483-4
UN: 2920
ADR: 8 II

Order Now

Specification Test Results

Assay (GC) 88.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR