2-Chloro-5-fluoropyrimidine 97%

Supplier: Thermo Scientific Chemicals
Danger

H55055-03 H55055-06
CAAAH55055-03EA 252.8 CAD
CAAAH55055-03 CAAAH55055-06 CAAAH55055-MD
2-Chloro-5-fluoropyrimidine 97%
2-Chloro-5-fluoropyrimidine

CAS: 62802-42-0 MDL: MFCD03788197

Formula: C₄H₂ClFN₂
Density: 1.073
Flash Pt: 65 °C(149 °F)
Storage Temperature: Ambient
MDL Number: MFCD03788197
CAS Number: 62802-42-0
UN: 1760
ADR: "8",III

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR