Benzylhydrazine dihydrochloride 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H54916-06
H54916-14
CAAAH54916-06EA
103.49
CAD
CAAAH54916-06
CAAAH54916-14
Benzylhydrazine dihydrochloride 97%
Benzylhydrazine dihydrochloride
Purity: 97%
CAS No.: 20570-96-1
Molecular Formula: C7H10N2.2HCl
Formula Weight: 195.09
Melting Point: 143-145°
Beilstein Reference: 3688990
UN No.: 2811
MDL No.: MFCD00012921
Formula:
C₇H₁₀N₂ Melting Pt: 143…145 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012921 CAS Number: 20570-96-1 EINECS: 243-887-1 ADR: 6.1,III |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...