4-Bromosalicylaldehyde 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H54850-03
H54850-06
CAAAH54850-03EA
255.96
CAD
CAAAH54850-03
CAAAH54850-06
CAAAH54850-MD
4-Bromosalicylaldehyde 97%
4-Bromosalicylaldehyde
CAS: 22532-62-3
MDL: MFCD06252606
Formula:
C₇H₅BrO₂ Melting Pt: 50…54 °C Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD06252606 CAS Number: 22532-62-3 UN: 3077 ADR: 9,III |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...