1-Methyl-1H-pyrazole-4-sulfonyl chloride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H54785-03
H54785-06
H54785-MD
CAAAH54785-03EA
237
CAD
CAAAH54785-03
CAAAH54785-06
CAAAH54785-MD
1-Methyl-1H-pyrazole-4-sulfonyl chloride 97%
1-Methyl-1H-pyrazole-4-sulfonyl chloride
Purity: 97%
CAS No.: 288148-34-5
Molecular Formula: C4H5ClN2O2S
Formula Weight: 179.98
Storage Temperature: -30°C to -10°C
UN No.: 3265
MDL No.: MFCD04968667
Formula:
C₄H₅ClN₂O₂S Storage Temperature: Refrigerator |
MDL Number:
MFCD04968667 CAS Number: 288148-34-5 UN: 3265 ADR: "8",III |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...