Coumarin-6-sulfonyl chloride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H54767-03
H54767-06
H54767-14
CAAAH54767-03EA
83.74
CAD
CAAAH54767-03
CAAAH54767-06
CAAAH54767-14
Coumarin-6-sulfonyl chloride 97%
Coumarin-6-sulfonyl chloride
Purity: 97%
CAS No.: 10543-42-7
Molecular Formula: C9H5ClO4S
Formula Weight: 244.65
Beilstein Reference: 196167
UN No.: 3261
MDL No.: MFCD01941320
Formula:
C₉H₅ClO₄S Storage Temperature: Ambient |
MDL Number:
MFCD01941320 CAS Number: 10543-42-7 UN: 3261 ADR: "8",III |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...