1,3-benzodioxole-5-sulfonyl chloride 95%

Supplier: Thermo Scientific Chemicals
Danger

H54755-03 H54755-06 H54755-MD
CAAAH54755-03EA 276.5 CAD
CAAAH54755-03 CAAAH54755-06 CAAAH54755-MD
1,3-benzodioxole-5-sulfonyl chloride 95%
1,3-benzodioxole-5-sulfonyl chloride

Purity: 95% CAS No.: 115010-10-1 Molecular Formula: C7H5ClO4S Formula Weight: 220.63 UN No.: 3261 MDL No.: MFCD04974524

Formula: C₇H₅ClO₄S
Storage Temperature: Ambient
MDL Number: MFCD04974524
CAS Number: 115010-10-1
UN: 3261
ADR: "8",III

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR