1H-imidazole-4-sulfonyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

H54642-03 H54642-06 H54642-14
CAAAH54642-03EA 142.67 CAD
CAAAH54642-03 CAAAH54642-06 CAAAH54642-14
1H-imidazole-4-sulfonyl chloride 97%
1H-imidazole-4-sulfonyl chloride

Purity: 97% CAS No.: 58767-51-4 Molecular Formula: C3H3ClN2O2S Formula Weight: 165.96 UN No.: 3261 MDL No.: MFCD08687938

Formula: C₃H₃ClN₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD08687938
CAS Number: 58767-51-4
UN: 3261
ADR: "8",III

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR