2,4-Dimethylthiazole-5-sulfonyl chloride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H54578-03
H54578-06
H54578-MD
CAAAH54578-03EA
162.74
CAD
CAAAH54578-03
CAAAH54578-06
CAAAH54578-MD
2,4-Dimethylthiazole-5-sulfonyl chloride 97%
2,4-Dimethylthiazole-5-sulfonyl chloride
Purity: 97%
CAS No.: 80466-80-4
Molecular Formula: C5H6ClNO2S2
Formula Weight: 211.68
Boiling Point: 82-84°/0.3mm
UN No.: 3265
MDL No.: MFCD00052746
Formula:
C₅H₆ClNO₂S₂ MW: 211.678 g/mol Boiling Pt: 82…84 °C/0.3 mm Storage Temperature: Ambient |
MDL Number:
MFCD00052746 CAS Number: 80466-80-4 UN: 3265 ADR: "8",III |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...