1-Boc-(2-aminomethyl)piperidine 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-(Aminomethyl)-1-piperidinecarboxylic acid tert-Butyl Ester, 2-Aminomethylpiperidine-1-carboxylic acid tert-butyl ester, 1-Boc-2-aminomethylpiperidine, 2-(Aminomethyl)-1-Boc-piperidine, 2-(Aminomethyl)-1-tert-butoxycarbonylpiperidine, 1-Boc-2-(Aminomethyl)piperidine, 1,1-Dimethylethyl 2-(aminomethyl)-1-piperidinecarboxylate, 2-(Aminomethyl)-1-piperidinecarboxylic acid tert-Butyl ester, tert-Butyl 2-(Aminomethyl)-1-piperidinecarboxylate, 2-Aminomethyl-1-Boc-piperidine
Purity: 95%
CAS No.: 370069-31-1
Molecular Formula: C11H22N2O2
Formula Weight: 214.31
Flash Point:
Formula:
C11H22N2O2 MW: 214.31 g/mol Density: 1.012 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD01317791 CAS Number: 370069-31-1 UN: 3082 ADR: 9 III |
Specification Test Results
Assay Minimum | 95 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...