indane-5-sulfonyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,3-Dihydro-1H-indene-5-sulfonyl chloride, 2,3-Dihydro-1H-indene-5-sulphonyl chloride

H54462-03 H54462-06 H54462-MD
CAAAH54462-03EA 147.26 CAD
CAAAH54462-03 CAAAH54462-06 CAAAH54462-MD
indane-5-sulfonyl chloride 97%
indane-5-sulfonyl chloride

Purity: 97% CAS No.: 52205-85-3 Molecular Formula: C9H9ClO2S Formula Weight: 216.69 UN No.: 3261 MDL No.: MFCD05237217

Formula: C₉H₉ClO₂S
MW: 216.679 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05237217
CAS Number: 52205-85-3
UN: 3261
ADR: "8",III

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR