2,1,3-benzoxadiazole-4-sulfonyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Benzofurazan-4-sulphonyl chloride, Benzofurazan-4-sulfonyl chloride

H54275-03 H54275-06 H54275-MD
CAAAH54275-03EA 316 CAD
CAAAH54275-03 CAAAH54275-06 CAAAH54275-MD
2,1,3-benzoxadiazole-4-sulfonyl chloride 97%
2,1,3-benzoxadiazole-4-sulfonyl chloride

Purity: 97% CAS No.: 114322-14-4 Molecular Formula: C6H3ClN2O3S Formula Weight: 218.61 UN No.: 3261 MDL No.: MFCD00068072

Formula: C₆H₃ClN₂O₃S
MW: 218.611 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00068072
CAS Number: 114322-14-4
UN: 3261
ADR: "8",III

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR