1,3-Dioxane 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: m-Dioxane, 1,3-Dioxacyclohexane

H54107-06 H54107-14
CAAAH54107-06EA 91.01 CAD
CAAAH54107-06 CAAAH54107-14
1,3-Dioxane 98%
1,3-Dioxane

CAS: 505-22-6 MDL: MFCD00006566

Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 105…106 °C
Melting Pt: –45 °C
Density: 1.032
Flash Pt: 5 °C (41 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006566
CAS Number: 505-22-6
EINECS: 208-005-1
UN: 1165
ADR: 3,II

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR