4'-(Methylsulfonyl)acetophenone 97+%

Supplier: Thermo Scientific Chemicals

Synonyms: 4'-(Methylsulphonyl)acetophenone, 4'-(Methylsulfonyl)acetophenone

H54072-03 H54072-06
CAAAH54072-03EA 54.35 CAD
CAAAH54072-03 CAAAH54072-06
4'-(Methylsulfonyl)acetophenone 97+%
4'-(Methylsulphonyl)acetophenone

CAS: 10297-73-1 MDL: MFCD00025072

Formula: C₉H₁₀O₃S
MW: 198.24 g/mol
Melting Pt: 126…129 °C
Storage Temperature: Ambient
MDL Number: MFCD00025072
CAS Number: 10297-73-1
EINECS: 233-672-0

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR