3,5-Dimethyl-1H-pyrazole-4-sulfonyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

H54050-03 H54050-06
CAAAH54050-03EA 146.47 CAD
CAAAH54050-03 CAAAH54050-06 CAAAH54050-MD
3,5-Dimethyl-1H-pyrazole-4-sulfonyl chloride 97%
3,5-Dimethyl-1H-pyrazole-4-sulfonyl chloride

Purity: 97% CAS No.: 80466-78-0 Molecular Formula: C5H7ClN2O2S Formula Weight: 194.64 UN No.: 3261 MDL No.: MFCD04969910

Formula: C₅H₇ClN₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD04969910
CAS Number: 80466-78-0
UN: 3261
ADR: "8",III

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR