4-(Trifluoromethyl)styrene 98% stabilized

Supplier: Thermo Scientific Chemicals
Warning

H53513-03 H53513-06
CAAAH53513-03EA 211.72 CAD
CAAAH53513-03 CAAAH53513-06
4-(Trifluoromethyl)styrene 98% stabilized
4-(Trifluoromethyl)styrene

Purity: 98% CAS No.: 402-50-6 Molecular Formula: C9H7F3 Formula Weight: 172.15 Storage Temperature: 2°C to 8°C Flash Point: 42°(108°F) Refractive Index: 1.466 Boiling Point: 65-66°/40mm Beilstein Reference: 1863548 UN No.: 1993 MDL No.: MFCD00075539

Formula: C₉H₇F₃
Boiling Pt: 65…66 °C/40 mm
Density: 1.165
Flash Pt: 42 °C(108 °F)
Storage Temperature: Ambient
MDL Number: MFCD00075539
CAS Number: 402-50-6
UN: 1993
ADR: "3",III

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR