3,4-Dimethylhexane 97%

Supplier: Thermo Scientific Chemicals
Danger

H53498-06 H53498-14
CAAAH53498-06EA 116.76 CAD
CAAAH53498-06 CAAAH53498-14
3,4-Dimethylhexane 97%
3,4-Dimethylhexane

CAS: 583-48-2 MDL: MFCD00039927

Formula: C₈H₁₈
MW: 114.23 g/mol
Boiling Pt: 119 °C
Melting Pt: -64
Density: 0.72
Flash Pt: 6 °C (43 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039927
CAS Number: 583-48-2
EINECS: 209-504-7
UN: 1262
ADR: 3 II

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR