Cyclopropyl(trimethylsilyl)acetylene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H53487-06
H53487-14
CAAAH53487-06EA
170.64
CAD
CAAAH53487-06
CAAAH53487-14
Cyclopropyl(trimethylsilyl)acetylene 97%
Cyclopropyl(trimethylsilyl)acetylene
Purity: 97%
CAS No.: 81166-84-9
Molecular Formula: C8H14Si
Formula Weight: 138.28
Flash Point: 25°(77°F)
Refractive Index: 1.454
Boiling Point: 83°/80mm
UN No.: 1993
MDL No.: MFCD00671355
Formula:
C₈H₁₄Si Boiling Pt: 83 °C/80 mm Flash Pt: 25 °C(77 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00671355 CAS Number: 81166-84-9 UN: 1993 ADR: "3",III |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...