2-Hexene cis- and trans-mixture tech. 85%, Technical Grade

Supplier: Thermo Scientific Chemicals
Danger

H53472-06 H53472-14
CAAAH53472-06EA 118.18 CAD
CAAAH53472-06 CAAAH53472-14
2-Hexene cis- and trans-mixture tech. 85%, Technical Grade
2-Hexene

CAS: 592-43-8 MDL: MFCD00009473

Formula: C₆H₁₂
MW: 84.16 g/mol
Boiling Pt: 67…69 °C
Melting Pt: –98.5 °C
Density: 0.678
Flash Pt: –20 °C (–4 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009473
CAS Number: 592-43-8
EINECS: 209-755-2
UN: 3295
ADR: 3,II

Order Now

Specification Test Results

Assay Minimum 85

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR