1-(Trimethylsilyl)-1-pentyne 98%

Supplier: Thermo Scientific Chemicals
Warning

H53423-06 H53423-14 H53423-22
CAAAH53423-06EA 52.93 CAD
CAAAH53423-06 CAAAH53423-14 CAAAH53423-22
1-(Trimethylsilyl)-1-pentyne 98%
1-(Trimethylsilyl)-1-pentyne

CAS: 18270-17-2 MDL: MFCD00077890

Formula: C₈H₁₆Si
Boiling Pt: 135 °C
Density: 0.765
Flash Pt: 23 °C(73 °F)
Storage Temperature: Ambient
MDL Number: MFCD00077890
CAS Number: 18270-17-2
UN: 1993
ADR: "3",III

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR