Ethyl-8-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate 95%

Supplier: Thermo Scientific Chemicals
Warning

H35916-03 H35916-06
CAAAH35916-03EA 96.06 CAD
CAAAH35916-03 CAAAH35916-06
Ethyl-8-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate 95%
Ethyl 8-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate

CAS: 353258-31-8 MDL: MFCD01833013

Formula: C₁₁H₈ClF₃N₂O₂
Melting Pt: 132…136 °C
Storage Temperature: Ambient
MDL Number: MFCD01833013
CAS Number: 353258-31-8

Order Now

Specification Test Results

Assay Minimum 95


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR