imidazo[1,2-a]pyridine-6-carbonitrile 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H35854-03
H35854-06
CAAAH35854-03EA
165.9
CAD
CAAAH35854-03
CAAAH35854-06
imidazo[1,2-a]pyridine-6-carbonitrile 95%
imidazo[1,2-a]pyridine-6-carbonitrile
Purity: 95%
CAS No.: 106850-34-4
Molecular Formula: C8H5N3
Formula Weight: 148.15
Melting Point: 161-163°
MDL No.: MFCD06659603
Formula:
C₈H₅N₃ Melting Pt: 161…163 °C Storage Temperature: Ambient |
MDL Number:
MFCD06659603 CAS Number: 106850-34-4 |
Specification Test Results
Assay Minimum | 95 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...