Pyrazolo[1,5-a]pyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

H35502-03 H35502-MD
CAAAH35502-03EA 224.36 CAD
CAAAH35502-03 CAAAH35502-MD
Pyrazolo[1,5-a]pyridine 97%
Pyrazolo[1,5-a]pyridine

CAS: 274-56-6 MDL: MFCD08752622

Formula: C₇H₆N₂
MW: 118.14 g/mol
Boiling Pt: 37…39 °C/0.2 mm
Storage Temperature: Ambient
MDL Number: MFCD08752622
CAS Number: 274-56-6

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR