3-Bromoimidazo[1,2-a]pyridine-6-carbonitrile 95%

Supplier: Thermo Scientific Chemicals
Warning

H35421-03 H35421-MD
CAAAH35421-03EA 141.57 CAD
CAAAH35421-03 CAAAH35421-MD
3-Bromoimidazo[1,2-a]pyridine-6-carbonitrile 95%
3-Bromoimidazo[1,2-a]pyridine-6-carbonitrile

CAS: 885950-21-0 MDL: MFCD06659617

Formula: C₈H₄BrN₃
Melting Pt: 207…209 °C
Storage Temperature: Ambient
MDL Number: MFCD06659617
CAS Number: 885950-21-0

Order Now

Specification Test Results

Assay Minimum 95


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR