4,5,6,7-Tetrahydrobenzo[b]thiophene-2-sulfonyl chloride 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H34470-03
H34470-MD
CAAAH34470-03EA
477.16
CAD
CAAAH34470-03
CAAAH34470-MD
4,5,6,7-Tetrahydrobenzo[b]thiophene-2-sulfonyl chloride 95%
4,5,6,7-Tetrahydrobenzo[b]thiophene-2-sulfonyl chloride
Purity: 95%
CAS No.: 128852-17-5
Molecular Formula: C8H9ClO2S2
Formula Weight: 236.74
Storage Temperature: -30°C to -10°C
Melting Point: 64-66°
UN No.: 3261
MDL No.: MFCD18825753
Formula:
C₈H₉ClO₂S₂ Melting Pt: 64…66 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD18825753 CAS Number: 128852-17-5 UN: 3261 ADR: "8",III |
Specification Test Results
Assay Minimum | 95 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...