4'-Methylbiphenyl-4-carboxylic acid 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4'-Methyl-4-biphenylcarboxylic acid

H34459-03 H34459-06
CAAAH34459-03EA 112.5 CAD
CAAAH34459-03 CAAAH34459-06
4'-Methylbiphenyl-4-carboxylic acid 96%
4'-Methylbiphenyl-4-carboxylic acid

CAS: 720-73-0 MDL: MFCD00448778

Formula: C₁₄H₁₂O₂
MW: 212.25 g/mol
Melting Pt: 257…258 °C
Storage Temperature: Ambient
MDL Number: MFCD00448778
CAS Number: 720-73-0

Order Now

Specification Test Results

Assay Minimum 96


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR