4-(2-Oxo-1-pyrrolidinyl)benzenesulfonyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-(2-Oxopyrrolidin-1-yl)benzene-1-sulphonyl chloride, 4-(2-Oxopyrrolidin-1-yl)benzene-1-sulfonyl chloride

H34441-03 H34441-06 H34441-MD
CAAAH34441-03EA 244.9 CAD
CAAAH34441-03 CAAAH34441-06 CAAAH34441-MD
4-(2-Oxo-1-pyrrolidinyl)benzenesulfonyl chloride 97%
4-(2-Oxo-1-pyrrolidinyl)benzenesulfonyl chloride

Purity: 97% CAS No.: 112539-09-0 Molecular Formula: C10H10ClNO3S Formula Weight: 259.71 Melting Point: 104-105° UN No.: 3261 MDL No.: MFCD00219329

Formula: C₁₀H₁₀ClNO₃S
MW: 259.704 g/mol
Melting Pt: 104…105 °C
Storage Temperature: Ambient
MDL Number: MFCD00219329
CAS Number: 112539-09-0
UN: 3261
ADR: "8",III

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR