1-(5-Trifluoromethyl-2-pyridyl)-1H-pyrazole-4-sulfonyl chloride 95%

Supplier: Thermo Scientific Chemicals
Danger

H34430-03 H34430-MD
CAAAH34430-03EA 122.92 CAD
CAAAH34430-03 CAAAH34430-06 CAAAH34430-MD
1-(5-Trifluoromethyl-2-pyridyl)-1H-pyrazole-4-sulfonyl chloride 95%
1-(5-Trifluoromethyl-2-pyridyl)-1H-pyrazole-4-sulfonyl chloride

Purity: 95% CAS No.: 1006441-36-6 Molecular Formula: C9H5ClF3N3O2S Formula Weight: 311.67 Melting Point: 87-89° UN No.: 3261 MDL No.: MFCD08689729

Formula: C₉H₅ClF₃N₃O₂S
MW: 311.663 g/mol
Melting Pt: 87…89 °C
Storage Temperature: Ambient
MDL Number: MFCD08689729
CAS Number: 1006441-36-6
UN: 3261
ADR: 8 III

Order Now

Specification Test Results

Assay Minimum 95

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR