7-Bromo-5-methylbenzo[b]furan 97%

Supplier: Thermo Scientific Chemicals
Warning

H34347-03 H34347-MD
CAAAH34347-03EA 195.92 CAD
CAAAH34347-03 CAAAH34347-MD
7-Bromo-5-methylbenzo[b]furan 97%
7-Bromo-5-methylbenzo[b]furan

Purity: 97% CAS No.: 35700-48-2 Molecular Formula: C9H7BrO Formula Weight: 211.06 MDL No.: MFCD11877828

Formula: C₉H₇BrO
Storage Temperature: Ambient
MDL Number: MFCD11877828
CAS Number: 35700-48-2

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR