1H-Indene-3-carboxylic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H34344-03
H34344-06
CAAAH34344-03EA
87.53
CAD
CAAAH34344-03
CAAAH34344-06
1H-Indene-3-carboxylic acid 97%
1H-Indene-3-carboxylic acid
Purity: 97%
CAS No.: 5020-21-3
Molecular Formula: C10H8O2
Formula Weight: 160.17
Melting Point: 161-165°
MDL No.: MFCD00086207
Formula:
C₁₀H₈O₂ Melting Pt: 161…165 °C Storage Temperature: Ambient |
MDL Number:
MFCD00086207 CAS Number: 5020-21-3 |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...