Pyridazine-3-carboxylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

H34341-03 H34341-06
CAAAH34341-03EA 97.8 CAD
CAAAH34341-03 CAAAH34341-06
Pyridazine-3-carboxylic acid 97%
Pyridazine-3-carboxylic acid

CAS: 2164-61-6 MDL: MFCD01646333

Formula: C₅H₄N₂O₂
Melting Pt: ca 195 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD01646333
CAS Number: 2164-61-6

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR