4,5,6,7-Tetrahydrobenzo[b]thiophene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H34309-03
H34309-06
CAAAH34309-03EA
233.84
CAD
CAAAH34309-03
CAAAH34309-06
CAAAH34309-MD
4,5,6,7-Tetrahydrobenzo[b]thiophene 97%
4,5,6,7-Tetrahydrobenzo[b]thiophene
Purity: 97%
CAS No.: 13129-17-4
Molecular Formula: C8H10S
Formula Weight: 138.23
Refractive Index: 1.5580
Boiling Point: 55°/1mm
MDL No.: MFCD19442759
Formula:
C₈H₁₀S Boiling Pt: 55 °C/1 mm Storage Temperature: Ambient |
MDL Number:
MFCD19442759 CAS Number: 13129-17-4 |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...