2-Amino-3-(trifluoromethyl)pyridine 97%

Supplier: Thermo Scientific Chemicals
Danger

H34303-06 H34303-14
CAAAH34303-06EA 103.17 CAD
CAAAH34303-06 CAAAH34303-14
2-Amino-3-(trifluoromethyl)pyridine 97%
2-Amino-3-(trifluoromethyl)pyridine

CAS: 183610-70-0 MDL: MFCD04972538

Formula: C₆H₅F₃N₂
Melting Pt: 70…72 °C
Storage Temperature: Ambient
MDL Number: MFCD04972538
CAS Number: 183610-70-0
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR