4'-Methylbiphenyl-4-carboxaldehyde 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4'-Methyl-[1,1'-biphenyl]-4-carbaldehyde
H34249-03
H34249-MD
CAAAH34249-03EA
197.5
CAD
CAAAH34249-03
CAAAH34249-MD
4'-Methylbiphenyl-4-carboxaldehyde 96%
4'-Methylbiphenyl-4-carboxaldehyde
Purity: 96%
CAS No.: 36393-42-7
Molecular Formula: C14H12O
Formula Weight: 196.25
Melting Point: 108-113°
MDL No.: MFCD02258938
Formula:
C₁₄H₁₂O MW: 196.249 g/mol Melting Pt: 108…113 °C Storage Temperature: Ambient |
MDL Number:
MFCD02258938 CAS Number: 36393-42-7 |
Specification Test Results
Assay Minimum | 96 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...