2-Chloro-3-iodo-1,4-dimethoxybenzene 97%

Supplier: Thermo Scientific Chemicals
Warning

H34232-03 H34232-06
CAAAH34232-03EA 82.95 CAD
CAAAH34232-03 CAAAH34232-06
2-Chloro-3-iodo-1,4-dimethoxybenzene 97%
2-Chloro-3-iodo-1,4-dimethoxybenzene

Purity: 97% CAS No.: 1335243-55-4 Molecular Formula: C8H8ClIO2 Formula Weight: 298.51 Melting Point: 129-131° MDL No.: MFCD22380712

Formula: C₈H₈ClIO₂
Melting Pt: 129…131 °C
Storage Temperature: Ambient
MDL Number: MFCD22380712
CAS Number: 1335243-55-4

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR