2'-Fluoro-4-biphenylmethanol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H34211-03
H34211-MD
CAAAH34211-03EA
206.98
CAD
CAAAH34211-03
CAAAH34211-MD
2'-Fluoro-4-biphenylmethanol 97%
2'-Fluoro-4-biphenylmethanol
Purity: 97%
CAS No.: 773872-57-4
Molecular Formula: C13H11FO
Formula Weight: 202.23
Melting Point: 57-59°
MDL No.: MFCD05980834
Formula:
C₁₃H₁₁FO Melting Pt: 57…59 °C Storage Temperature: Ambient |
MDL Number:
MFCD05980834 CAS Number: 773872-57-4 |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...